EMAIL THIS PAGE TO A FRIEND
The Korean journal of parasitology
Hypersensitive reaction to praziquantel in a clonorchiasis patient.
PMID 22072827
Abstract
Praziquantel is the drug of choice for clonorchiasis. Since clonorchiasis is endemic in most river basins, praziquantel has been widely used for 30 years in Korea. A 54-year-old Korean woman suffered from hypersensitive reactions, such as nausea, dyspnea, rash, and urticaria after taking the first dose of praziquantel to treat clonorchiasis. She ingested one dose again and the same symptoms appeared, and she was treated at a clinic with anti-histamines. She tried one more dose with anti-histamines but found the same symptoms. Later, she was found to pass eggs of Clonorchis sinensis and medicated with flubendazole. The hypersensitive reaction to praziquantel is rare but occurs. This is the 5th case report in the world.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34091
Flubendazol, VETRANAL, analytical standard
C16H12FN3O3