EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Base-mediated stereospecific synthesis of aryloxy and amino substituted ethyl acrylates.
PMID 22073965
Abstract
The stereospecific synthesis of aryloxy and amino substituted E- and Z-ethyl-3-acrylates is of interest because of their potential in the polymer industry and in medicinal chemistry. During work on a copper-catalyzed cross-coupling reaction of ethyl (E)- and (Z)-3-iodoacrylates with phenols and N-heterocycles, we discovered a very simple (nonmetallic) method for the stereospecific synthesis of aryloxy and amino substituted acrylates. To study this long-standing problem on the stereoselectivity of aryloxy and amino substituted acrylates, a series of O- and N-substituted nucleophiles was allowed to react with ethyl (E)- and (Z)-3-iodoacrylates. Screening of different bases indicated that DABCO (1,4-diazabicyclo[2.2.2]octane) afforded successful conversion of ethyl (E)- and (Z)-3-iodoacrylates into aryloxy and amino substituted ethyl acrylates in a stereospecific manner. Herein are the details of this DABCO-mediated stereospecific synthesis of aryloxy and amino substituted E- or Z-acrylates.