EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Longitudinal zipping/unzipping of self-assembled organic tubes.
PMID 22073969
Abstract
Stimuli-responsive organic tubes are an attractive supramolecular assembly which has potential applications as a controlled release vehicle. We synthesize a smart organic tube by the coassembly of lithocholic acid (LCA) and taurolithocholic acid (TLCA) in aqueous solution. The coassembled LCA/TLCA tubes can be longitudinally unzipped into flat sheets by capillary force after being dehydrated on substrates. Consequently, the encapsulated guest molecules are released from the unzipping tubes. After the release of guest molecules, the flat sheets can be zipped back into hollow tubes upon hydration with aqueous solution. The zipping/unzipping LCA/TLCA tubes provide a new type of delivery vehicles, which may have potential for surface decontaminations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7515
Sodium taurolithocholate
C26H44NNaO5S