EMAIL THIS PAGE TO A FRIEND
Current radiopharmaceuticals
Visualising neuroinflammation in post-stroke patients: a comparative PET study with the TSPO molecular imaging biomarkers [11C]PK11195 and [11C]vinpocetine.
PMID 22074478
Abstract
With the main objective of comparing the prospective diagnostic power of two 11C-labelled molecular imaging biomarkers with affinity for TSPO and used for the visualisation of activated microglia after a stroke, we measured with positron emission tomography (PET) in four post-stroke patients the regional brain uptake and binding potential of [11C]vinpocetine and [11C]PK11195. Percentage standard uptake values (%SUV) and binding potential (BPND) were used as outcome measures. The total peak brain uptake value and average global brain uptake value were higher for [11C]vinpocetine than for [11C]PK11195. The regional %SUV values were significantly higher for [11C]vinpocetine than for [11C]PK11195 in the hemispheres as well as in almost all standard brain regions. The %SUV values of [11C]vinpocetine were higher in the peri-infarct zone than in the ischaemic core, however, the difference did not prove to be significant. There was basically no difference in %SUV values between the ischaemic core and the peri-infarct zone for [11C]PK11195. The BPND values for [11C]vinpocetine were higher in all standard regions than those for [11C]PK11195, but the difference was not significant between them. The BPND values of [11C]vinpocetine were higher in the peri-infarct zone than in the ischaemic core, however, the difference did not prove to be significant. A comparative analysis of the two ligands indicates that [11C]vinpocetine shows a number of favourable characteristics over [11C]PK11195, but to demonstrate that it may serve as a prospective molecular imaging biomarker of microglia activation in post-stroke patients, further studies are required.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V6383
Vinpocetine, ≥98%, solid
C22H26N2O2