EMAIL THIS PAGE TO A FRIEND
Journal of pediatric urology
Commentary to "Intermediate-long term follow-up indicates low risk of recurrence after double HIT endoscopic treatment for primary vesicoureteral reflux (VUR)".
PMID 22075263
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000744 Dextranomer, European Pharmacopoeia (EP) Reference Standard