EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A hybrid biocathode: surface display of O2-reducing enzymes for microbial fuel cell applications.
PMID 22075939
Abstract
Laccase and bilirubin oxidase were successfully displayed on the surface of yeast cells. Subsequently, these modified yeast cells were used in the cathode compartment of a microbial fuel cell. The performance of the fuel cells is compared.
Related Materials