EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Geranylgeranylacetone protects against myocardial ischemia and reperfusion injury by inhibiting high-mobility group box 1 protein in rats.
PMID 22075975
Abstract
The high mobility group box 1 (HMGB1) protein plays an important role in myocardial ischemia and reperfusion (I/R) injury. Geranylgeranylacetone (GGA), a heat shock protein 72 inducer, has been reported to reduce myocardial I/R injury. The aim of this study was to investigate the cardioprotective mechanism of GGA during myocardial I/R injury in rats. Anesthetized male rats were treated once with GGA (200 mg/kg, p.o.) 24 h before ischemia, and subjected to ischemia for 30 min, followed by reperfusion for 4 h. Lactate dehydrogenase (LDH), creatine kinase (CK), malondialdehyde (MDA), superoxide dismutase (SOD) activity and infarct size were measured. HMGB1 expression was assessed by immunoblotting. The results showed that pre-treatment with GGA (200 mg/kg) significantly reduced the infarct size and the levels of LDH and CK after 4 h of reperfusion (all P<0.05). GGA also significantly inhibited the increase in MDA levels and the decrease in SOD levels (both P<0.05). Meanwhile, GGA considerably suppressed the expression of HMGB1 induced by I/R. The present study suggests that GGA is capable of attenuating myocardial I/R injury by inhibiting HMGB1 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5048
Geranylgeranylacetone
C23H38O