EMAIL THIS PAGE TO A FRIEND
Cytokine
Interleukin-13 enhanced Ca2+ oscillations in airway smooth muscle cells.
PMID 22078634
Abstract
Physiological mechanisms associated with interleukin-13 (IL-13), a key cytokine in asthma, in intracellular Ca(2+) signaling in airway smooth muscle cells (ASMCs) remain unclear. The aim of this study was to assess effects of IL-13 on Ca(2+) oscillations in response to leukotriene D4 (LTD4) in human cultured ASMCs. LTD4-induced Ca(2+) oscillations in ASMCs pretreated with IL-13 were imaged by confocal microscopy. mRNA expressions of cysteinyl leukotriene 1 receptors (CysLT1R), CD38, involved with the ryanodine receptors (RyR) system, and transient receptor potential canonical (TRPC), involved with store-operated Ca(2+) entry (SOCE), were determined by real-time PCR. In IL-13-pretreated ASMCs, frequency of LTD4-induced Ca(2+) oscillations and number of oscillating cells were significantly increased compared with untreated ASMCs. Both xestospongin C, a specific inhibitor of inositol 1,4,5-triphosphate receptors (IP(3)R), and ryanodine or ruthenium red, inhibitors of RyR, partially blocked LTD4-induced Ca(2+) oscillations. Ca(2+) oscillations were almost completely inhibited by 50 μM of 2-aminoethoxydiphenyl borate (2-APB), which dominantly blocks SOCE but not IP(3)R at this concentration. Pretreatment with IL-13 increased the mRNA expressions of CysLT1R and CD38, but not of TRPC1 and TRPC3. We conclude that IL-13 enhances frequency of LTD4-induced Ca(2+) oscillations in human ASMCs, which may be cooperatively modulated by IP(3)R, RyR systems and possibly by SOCE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5011
Leukotriene D4, ~50 μg/mL (in methanol/ammonium acetate buffer, 70:30, pH 5.6), ≥97%
C25H40N2O6S