EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
The palladium-catalyzed desulfitative cyanation of arenesulfonyl chlorides and sodium sulfinates.
PMID 22080409
Abstract
A palladium-catalyzed desulfitative cyanation of arenesulfonyl chlorides and sodium sulfinates has been developed, providing aryl nitriles in moderate to excellent yields. It represents a facile procedure to access aryl nitriles.
Related Materials