EMAIL THIS PAGE TO A FRIEND
Nature immunology
Harnessing of the nucleosome-remodeling-deacetylase complex controls lymphocyte development and prevents leukemogenesis.
PMID 22080921
Abstract
Cell fate depends on the interplay between chromatin regulators and transcription factors. Here we show that activity of the Mi-2β nucleosome-remodeling and histone-deacetylase (NuRD) complex was controlled by the Ikaros family of lymphoid lineage-determining proteins. Ikaros, an integral component of the NuRD complex in lymphocytes, tethered this complex to active genes encoding molecules involved in lymphoid differentiation. Loss of Ikaros DNA-binding activity caused a local increase in chromatin remodeling and histone deacetylation and suppression of lymphoid cell-specific gene expression. Without Ikaros, the NuRD complex also redistributed to transcriptionally poised genes that were not targets of Ikaros (encoding molecules involved in proliferation and metabolism), which induced their reactivation. Thus, release of NuRD from Ikaros regulation blocks lymphocyte maturation and mediates progression to a leukemic state by engaging functionally opposing epigenetic and genetic networks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
MABE911
Anti-Aiolos Antibody, clone 9D10, This Anti-Aiolos Antibody, clone 9D10 is validated for use in Western Blotting and ChIP-seq and Chromatin Immunoprecipitation (ChIP) for the detection of Aiolos.
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
MABE912
Anti-Ikaros Antibody, clone 4E9, This Anti-Ikaros Antibody, clone 4E9 is validated for use in Western Blotting and Flow Cytometry and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Ikaros.
MABE913
Anti-Ikaros Antibody, clone Ik14, Anti-Ikaros, clone Ik14 is an antibody against Ikaros for use in WB, ChIP, ChIP-seq.
MABE914
Anti-Mi-2β Antibody, clone 16G4, This Anti-Mi-2β Antibody, clone 16G4 is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Mi-2β.
MABE920
Anti-Mi-2β Antibody, clone 17H11, This Anti-Mi-2β Antibody, clone 17H11 is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Mi-2β.
MABE915
Anti-Mi-2β Antibody, clone 2G8, This Anti-Mi-2β Antibody, clone 2G8 is validated for use in Western Blotting and ChIP-seq and Culture Confirmation for the detection of Mi-2β.
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ICC, IP.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.