EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Bensingtonia rectispora sp. nov. and Bensingtonia bomiensis sp. nov., ballistoconidium-forming yeast species from Tibetan plant leaves.
PMID 22081720
Abstract
Five yeast strains isolated from plant leaves collected in south-east Tibet formed cream to brownish colonies and produced asymmetrical ballistoconidia and CoQ-9 as the major ubiquinone. Sequence analysis of the 26S rRNA D1/D2 domain and the internal transcribed spacer region indicated that these strains represented two novel species of the genus Bensingtonia. The names Bensingtonia rectispora sp. nov. (type strain XZ 4C5(T) = CGMCC 2.02635(T) = CBS 10710(T)) and Bensingtonia bomiensis sp. nov. (type strain XZ 33D1(T) = CGMCC 2.02670(T) = CBS 10713(T)) are proposed for the two novel species, which are phylogenetically closely related to Bensingtonia naganoensis, Bensingtonia pseudonaganoensis and the type species of the genus, Bensingtonia ciliata.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27597
Coenzyme Q9, ≥96.0% (HPLC)
C54H82O4