EMAIL THIS PAGE TO A FRIEND
Addiction (Abingdon, England)
Double-blind placebo-controlled evaluation of the PROMETA™ protocol for methamphetamine dependence.
PMID 22082089
Abstract
To evaluate the efficacy and safety of the PROMETA™ Protocol for treating methamphetamine dependence. A double-blind, placebo-controlled 108-day study with random assignment to one of two study conditions: active medication with flumazenil (2 mg infusions on days 1, 2, 3, 22, 23), gabapentin (1200 mg to day 40) and hydroxazine (50 mg to day 10) versus placebo medication (with active hydroxazine only). Three substance abuse treatment clinics: two in-patient, one out-patient. Treatment-seeking, methamphetamine-dependent adults (n = 120). Primary outcome was percentage of urine samples testing negative for methamphetamine during the trial. No statistically significant between-group differences were detected in urine drug test results, craving, treatment retention or adverse events. The PROMETA protocol, consisting of flumazenil, gabapentin and hydroxyzine, appears to be no more effective than placebo in reducing methamphetamine use, retaining patients in treatment or reducing methamphetamine craving.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8885
Hydroxyzine dihydrochloride, ≥98%
C21H27ClN2O2 · 2HCl
H9010
Hydroxyzine pamoate salt, analytical standard
C21H27ClN2O2 · C23H16O6