EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Suzuki-Miyaura coupling of heteroaryl boronic acids and vinyl chlorides.
PMID 22083103
Abstract
A protocol for the Suzuki-Miyaura coupling of heteroaryl boronic acids and vinyl chlorides that minimizes protodeboronation is described. A combination of catalytic amounts of Pd(OAc)(2) and SPhos in conjunction with CsF in isopropanol effectively affords a variety of coupled products. Surprisingly, a dramatic temperature dependence in product selectivity was observed.