EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Barcoded nucleotides.
PMID 22083884
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258555
p-Phenylene diisothiocyanate, 98%
C8H4N2S2