EMAIL THIS PAGE TO A FRIEND
Zhonghua bing li xue za zhi = Chinese journal of pathology
[Compare three methods to detect the Pneumocystis carinii in the bronchoalveolar wash sample of AIDS patients].
PMID 22088377