EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Unidirectional growth of benzil crystal from solution by Sankaranarayanan-Ramasamy method and its characterization.
PMID 22088560
Abstract
Good quality <100> benzil single crystal with a diameter 18 mm and length 75 mm was successfully grown from solution by the unidirectional growth method of Sankaranarayanan-Ramasamy (SR) for the first time in the literature. The seed crystals have been harvested from conventional solution growth technique and subsequently used for unidirectional growth. The grown crystal was subjected to various characterization studies. The results of UV-vis spectral analysis, photoluminescence, etching and microhardness studies were compared with conventional solution grown crystal to that of SR method grown crystal. The quality of SR method grown benzil crystal is better than conventional solution grown crystal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5151
Benzil, 98%
C14H10O2
78600
Phenylglyoxal monohydrate, crystallized, ≥97.0% (GC)
C8H6O2 · H2O