EMAIL THIS PAGE TO A FRIEND
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases
Comparative evaluation of intratracheal colistimethate sodium, imipenem, and meropenem in BALB/c mice with carbapenem-resistant Acinetobacter baumannii pneumonia.
PMID 22088863
Abstract
The identification of the optimal agent for administration via the respiratory tract when treating pneumonia caused by carbapenem-resistant Acinetobacter baumannii (CRAB). A murine model of acute CRAB pneumonia was established by intratracheal (i.t.) inoculation with 2.5 × 10⁷ colony-forming units (CFU) of A. baumannii strain Ab396 plus 10% porcine mucin. After 4h the infected BALB/c mice were treated intratracheally with 25μl of either 0.85% saline (control group), colistimethate sodium (CMS) (166 666 U/kg, CMS group), imipenem/cilastatin (30/30 mg/kg, imipenem group), or meropenem (20mg/kg, meropenem group), every 8h. The therapeutic efficacy of these agents was examined. A. baumannii strain Ab396 was susceptible to CMS only. However, meropenem treatment did give a significantly superior survival rate (100%) compared to treatment with imipenem (50%), CMS (33%), or saline (0%) (p<0.001 vs. the control and CMS groups, p=0.006 vs. the imipenem group, by log-rank test). Furthermore, compared to the other groups, the meropenem group demonstrated significantly more favorable results in terms of tissue penetration of the antibiotic, bacterial clearance, normalization of the wet lung-to-body weight ratio, and down-regulation of pro-inflammatory cytokine levels in the lungs. Administration of meropenem via the respiratory tract proved to have the best therapeutic efficacy among the antibiotics tested when treating advanced murine CRAB pneumonia.