EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Studying the interactions of a platinum(II) 9-aminoacridine complex with proteins and oligonucleotides by ESI-TOF MS.
PMID 22089173
Abstract
The interaction of a novel Pt complex, [Pt(dmba)(N9-9AA)(PPh(3))](+)1 (dmba = N,N-dimethylbenzylamine-κN,κC; 9AA = 9-aminoacridine), which exhibits anti-tumor activity, with certain key proteins has been monitored by ESI-MS. Also, the interaction of 1 with a designed double-stranded oligonucleotide containing the GG motif has been followed by mass spectrometry as well as by fluorimetry. The results obtained show the low interaction of 1 with the considered proteins and the absence of covalent interaction with the oligonucleotides, but the fluorimetric data confirm the π-π interaction of 1 with the double-stranded DNA, which is probably the reason of the previously reported activity of 1 in several tumor cell lines.
Related Materials