EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Glycolate and 2-phosphoglycolate content of tissues measured by ion chromatography coupled to mass spectrometry.
PMID 22093610
Abstract
Glycolate and 2-phosphoglycolate (PG) are 2-carbon monocarboxylic acids with ill-defined metabolic roles. Their concentrations have not yet been described in tissues apart from body fluids and erythrocytes. We describe the use of ion chromatography coupled with mass spectrometry (IC-MS) to quantify levels of glycolate and PG in tissue. Sample preparation and analysis can be performed within an hour. Low concentrations of glycolate (12-48 nmol/g) and PG (4-17 nmol/g) were detected in all tissues. The availability of this IC-MS assay will facilitate investigations of the origin, function, and metabolism of glycolate and PG in tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72764
2-Phosphoglycolic acid lithium salt, ≥95.0% (HPLC)
C2H5O6P · xLi+