EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Effect of antibody modifications on its biomolecular binding as determined by surface plasmon resonance.
PMID 22093612
Abstract
A surface plasmon resonance (SPR)-based procedure was developed to determine the effect of antibody modifications on its biomolecular binding behavior. Mouse immunoglobulin G (IgG) was immobilized on a protein A-functionalized gold-coated SPR chip. Goat anti-mouse IgG and its various commercially available modifications (i.e., conjugated with atto 550, atto 647, tetramethylrhodamine isothiocyanate [TRITC], horseradish peroxidase [HRP], or biotin) were employed in exactly the same concentration for the detection of mouse IgG. The various modifications of goat anti-mouse IgG decreased its biomolecular binding to mouse IgG in the order of unmodified>HRP-labeled>atto 550-labeled>biotinylated>TRITC-labeled>atto 647-labeled.