EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Regio- and stereoselective synthesis of cyclopentenones: intermolecular pseudo-Pauson-Khand cyclization.
PMID 22095679
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C112909
2-Cyclopenten-1-one, 98%
C5H6O