EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
The marriage of organocatalysis with metal catalysis: access to multisubstituted chiral 2,5-dihydropyrroles by cascade iminium/enamine-metal cooperative catalysis.
PMID 22095848
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50900
Propargylamine, 98%
C3H5N
P50919
Propargylamine hydrochloride, 95%
C3H5N · HCl