EMAIL THIS PAGE TO A FRIEND
Liver international : official journal of the International Association for the Study of the Liver
Sphingosine kinase 1 promotes tumour cell migration and invasion via the S1P/EDG1 axis in hepatocellular carcinoma.
PMID 22098666
Abstract
Sphingosine kinase 1 (SphK1), which phosphorylates sphingosine to sphingosine-1-phosphate (S1P), is overexpressed in various types of cancers, and may act as an oncogene in tumorigenesis. However, little is known about the precise role of the SphK1/S1P pathway in human liver cancer, especially regarding the metastasis of hepatocellular carcinoma (HCC). The expression of SphK1 was detected by quantitative reverse-transcription PCR. In addition, transwell cell migration and invasion assay were carried out for functional analysis. Furthermore, the level of S1P was quantified by ELISA and Rac1/Cdc42 GTPase activation was assessed by western blot analysis. The levels of SphK1 mRNA are commonly up-regulated in HCC patients and human liver cancer cell migration and invasion can be promoted by the overexpression of SphK1. In addition, inhibition of SphK1 with either a SphK1 inhibitor or siRNA reduced human liver cancer cell migration and invasion. Furthermore, overexpression of SphK1 increased S1P levels, and the exogenous addition of S1P increased liver cell migration and invasion through the EDG1 receptor. The results from this study provide strong evidence of a role for the SphK1/S1P/EDG1 pathway in liver metastasis, thus making it an attractive therapeutic target for the development of new anti-HCC drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8326
SKI 5C, ≥98% (HPLC)
C26H45NO4