EMAIL THIS PAGE TO A FRIEND
Journal of natural medicines
New flav-3-en-3-ol glycosides, kaempferiaosides C and D, and acetophenone glycosides, kaempferiaosides E and F, from the rhizomes of Kaempferia parviflora.
PMID 22101441
Abstract
Two new flav-3-en-3-ol glycosides, kaempferiaosides C (3) and D(4), and two new acetophenone glycosides, kaempferiaosides E (5) and F (6), were isolated from the Thai natural medicine Krachai Dum, the rhizomes of Kaempferia parviflora Wall. ex Baker. Their structures were established mainly on the basis of 1D and 2D NMR spectral data.
Related Materials