EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Differential genotoxic effects of subchronic exposure to ethyl tertiary butyl ether in the livers of Aldh2 knockout and wild-type mice.
PMID 22102104
Abstract
Ethyl tertiary butyl ether (ETBE) is used as an additive to gasoline to reduce carbon monoxide emissions in some developed countries. So far, ETBE was not found with positive results in many genotoxic assays. This study is undertaken to investigate the modifying effects of deficiency of aldehyde dehydrogenase 2 (ALDH2) on the toxicity of ETBE in the livers of mice. Eight-week-old wild-type (WT) and Aldh2 knockout (KO) C57BL/6 mice of both sexes were exposed to 0, 500, 1,750, and 5,000 ppm ETBE for 6 h/day with 5 days per weeks for 13 weeks. Histopathology assessments and measurements of genetic effects in the livers were performed. Significantly increased accidences of centrilobular hypertrophy were observed in the livers of WT and KO mice of both sexes in 5,000 ppm group; there was a sex difference in centrilobular hypertrophy between male and female KO mice, with more severe damage in the males. In addition, DNA strand breaks, 8-hydroxyguanine DNA-glycosylase (hOGG1)-modified oxidative base modification, and 8-hydroxydeoxyguanosine as genetic damage endpoints were significantly increased in three exposure groups in KO male mice, while these genotoxic effects were only found in 5,000 ppm group of KO female mice. In WT mice, significant DNA damage was seen in 5,000 ppm group of male mice, but not in females. Thus, sex differences in DNA damage were found not only in KO mice, but also in WT mice. These results suggest that ALDH2 polymorphisms and sex should be taken into considerations in predicting human health effects of ETBE exposure.