EMAIL THIS PAGE TO A FRIEND
Journal of forensic sciences
Latent fingermark development on a range of porous substrates using ninhydrin analogs--a comparison with ninhydrin and 1,8-diazofluoren.
PMID 22103855
Abstract
Three relatively new reagents for developing latent fingermarks on porous substrates, 1,2-indandione (IND), 5-methylthioninhydrin (5-MTN), and lawsone, are compared with the more widely used ninhydrin and 1,8-diazofluoren (DFO). Developed latent fingermark visualization on 10 different substrates comprising colored papers, cardboard, and cellophane rather than conventional printer and writing/notepad paper is assessed using latent fingermark deposits from 48 donors. Results show improved fluorescent fingermark visualization using IND compared with DFO on a range of colored cardboards and thick white paper, thus extending the range of substrates known to yield improved visualization with IND. Adding zinc chloride to IND failed to yield any further improvement in fluorescent fingermark visualization. 5-MTN (with and without zinc chloride posttreatment) showed no improvement in visualization compared with ninhydrin and DFO although visible fingermarks were developed. Lawsone produced fluorescent visible fingermarks only with white substrates, which were inferior to those produced with DFO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H46805
2-Hydroxy-1,4-naphthoquinone, 97%
C10H6O3