EMAIL THIS PAGE TO A FRIEND
Journal of clinical lipidology
Letter to the editor.
PMID 22108154
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8281 Safflower seed oil from Carthamus tinctorius seed