EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Intermolecular [3+2] cycloaddition of cyclopropylamines with olefins by visible-light photocatalysis.
PMID 22109785
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125504
Cyclopropylamine, 98%
C3H7N