EMAIL THIS PAGE TO A FRIEND
Enzyme and microbial technology
Production of lactobionic acid and sorbitol from lactose/fructose substrate using GFOR/GL enzymes from Zymomonas mobilis cells: a kinetic study.
PMID 22112407
Abstract
In this work, we have investigated the kinetics of the biotechnological production of lactobionic acid (LBA) and sorbitol by the catalytic action of glucose-fructose oxidoreductase (GFOR) and glucono-δ-lactonase (GL) enzymes. The cells of bacterium Zymomonas mobilis ATCC 29191 containing this enzymatic complex were submitted to permeabilization and reticulation procedures. The effect of the concentration of substrates on the rate of product formation using a mobilized cell system was investigated. The application of higher fructose concentration seems to not affect the initial rate of formation of the bionic acid. Under conditions of low initial concentration of lactose, the experimental kinetic data of the bi-substrate reaction were modelled by assuming a rate equation of the classical ping-pong mechanism. The found kinetic parameters displayed a low affinity of the GFOR enzyme for both substrates. The enzymatic system did not exhibit normal Michaelis-Menten kinetics in response to a change of concentration of lactose, when fructose was held constant, presenting a sigmoid relationship between initial velocity and substrate concentration. A rate equation based on Hill kinetics was used to describe the kinetic behaviour of this enzyme-substituted reaction at higher lactose concentrations. The results from batch experiments using immobilized cells within Ca-alginate beads revealed that there is no pronounced occurrence of mass transfer limitations on LBA production for beads with 1.2 mm in average diameter. This discussion aids for defining the best operating conditions to maximize the productivity for LBA and sorbitol in this bioconversion and also for reducing the complexity of downstream separation processes.