EMAIL THIS PAGE TO A FRIEND
Journal of infection in developing countries
Mid-arm and epitrochlear lymphadenopathy: a clinico-radiological surprise.
PMID 22112738
Abstract
An 18-year-old man presented with multiple subcutaneous nodules over the anteromedial aspect of his right lower arm. Based on findings of sonography, histopathology, circulating filarial antigen test, and therapeutic response to diethylcarbazine, a diagnosis of filarial lymphadenopathy affecting mid-arm nodes and epitrochlear lymph nodes was made.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8765
Diethylcarbamazine citrate salt, ≥98%
C16H29N3O8