EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Approaching tumour therapy beyond platinum drugs: status of the art and perspectives of ruthenium drug candidates.
PMID 22112845
Abstract
The study of metal complexes for the treatment of cancer diseases has resulted in the identification of some unique properties of ruthenium-based compounds. Among these inorganic-based agents, two of them, namely the ruthenium(III) drugs NAMI-A and KP1019 have undertaken with some success the clinical evaluations of phase I and preliminary phase II trials in patients. Here we highlight the strategies that have led to the discovery of metal-based (NAMI-A and KP1019) and of organometallic (RM175, RAPTA-T, RDC11 and DW1/2) ruthenium-based complexes, and we report their main biological/pharmacological characteristics and expectations for further development.