EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Arbitrary self-assembly of peptide extracellular microscopic matrices.
PMID 22113878
Abstract
Two faces for one matrix: A single bifaceted cyclopeptide block forms highly branched, porous, and intricate fibrillar networks, which span microscopic dimensions and mimic the extracellular matrix to support cell growth and proliferation. The peptide block has two domains connected with triglycine linkers (GGG); the domains consist of positively (blue) and negatively (red) charged heptads that provide interactions between different blocks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1377
Gly-Gly-Gly
C6H11N3O4
50239
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
C6H11N3O4