EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Two new compounds from an endophytic fungus Pestalotiopsis heterocornis.
PMID 22115029
Abstract
Two new compounds, 7-hydroxy-5-methoxy-4,6-dimethyl-7-O-α-L-rhamnosyl-phthalide and 7-hydroxy-5-methoxy-4,6-dimethyl-7-O-β-D-glucopyranosyl-phthalide, along with one known and related metabolite 7-hydroxy-5-methoxy-4,6-dimethylphthalide were isolated from the EtOAc extract of fermentation broth of an endophytic fungus Pestalotiopsis heterocornis. The structures of these compounds were elucidated on the basis of spectroscopic methods (UV, IR, HR-ESI-MS, 1D NMR, and 2D NMR).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P39605
Phthalide, 98%
C8H6O2