EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Intensified chemotherapy with ACVBP plus rituximab versus standard CHOP plus rituximab for the treatment of diffuse large B-cell lymphoma (LNH03-2B): an open-label randomised phase 3 trial.
PMID 22118442
Abstract
The outcome of diffuse large B-cell lymphoma has been substantially improved by the addition of the anti-CD20 monoclonal antibody rituximab to chemotherapy regimens. We aimed to assess, in patients aged 18-59 years, the potential survival benefit provided by a dose-intensive immunochemotherapy regimen plus rituximab compared with standard treatment plus rituximab. We did an open-label randomised trial comparing dose-intensive rituximab, doxorubicin, cyclophosphamide, vindesine, bleomycin, and prednisone (R-ACVBP) with subsequent consolidation versus standard rituximab, doxorubicin, cyclophosphamide, vincristine, and prednisone (R-CHOP). Random assignment was done with a computer-assisted randomisation-allocation sequence with a block size of four. Patients were aged 18-59 years with untreated diffuse large B-cell lymphoma and an age-adjusted international prognostic index equal to 1. Our primary endpoint was event-free survival. Our analyses of efficacy and safety were of the intention-to-treat population. This study is registered with ClinicalTrials.gov, number NCT00140595. One patient withdrew consent before treatment and 54 did not complete treatment. After a median follow-up of 44 months, our 3-year estimate of event-free survival was 81% (95% CI 75-86) in the R-ACVBP group and 67% (59-73) in the R-CHOP group (hazard ratio [HR] 0·56, 95% CI 0·38-0·83; p=0·0035). 3-year estimates of progression-free survival (87% [95% CI, 81-91] vs 73% [66-79]; HR 0·48 [0·30-0·76]; p=0·0015) and overall survival (92% [87-95] vs 84% [77-89]; HR 0·44 [0·28-0·81]; p=0·0071) were also increased in the R-ACVBP group. 82 (42%) of 196 patients in the R-ACVBP group experienced a serious adverse event compared with 28 (15%) of 183 in the R-CHOP group. Grade 3-4 haematological toxic effects were more common in the R-ACVBP group, with a higher proportion of patients experiencing a febrile neutropenic episode (38% [75 of 196] vs 9% [16 of 183]). Compared with standard R-CHOP, intensified immunochemotherapy with R-ACVBP significantly improves survival of patients aged 18-59 years with diffuse large B-cell lymphoma with low-intermediate risk according to the International Prognostic Index. Haematological toxic effects of the intensive regimen were raised but manageable. Groupe d'Etudes des Lymphomes de l'Adulte and Amgen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4890
Vindesine sulfate salt hydrate, ≥95% (HPLC)
C43H55N5O7 · H2SO4 · xH2O