EMAIL THIS PAGE TO A FRIEND
Journal of postgraduate medicine
Atypical presentation of joint infection by an unusual organism.
PMID 22120866
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9393
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic
C19H17ClN3NaO5S · H2O