EMAIL THIS PAGE TO A FRIEND
Organic letters
Radical cyclization cascades of unsaturated Meldrum's acid derivatives.
PMID 22126403
Abstract
Unsaturated, differentially substituted Meldrum's acid derivatives undergo cascade cyclizations upon ester reduction with SmI(2)-H(2)O. The cascade cyclizations proceed in good yield and with high diastereocontrol and convert simple, achiral starting materials to complex molecular architectures, bearing up to four stereocenters, in a single operation. The cascades are triggered by the generation and trapping of unusual radical-anions formed by electron transfer to the ester carbonyl.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210145
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
C6H8O4