EMAIL THIS PAGE TO A FRIEND
Nucleus (Austin, Tex.)
Barrier-to-Autointegration Factor influences specific histone modifications.
PMID 22127260
Abstract
Defects in the nuclear envelope or nuclear 'lamina' networks cause disease and can perturb histone posttranslational (epigenetic) regulation. Barrier-to-Autointegration Factor (BAF) is an essential but enigmatic lamina component that binds lamins, LEM-domain proteins, DNA and histone H3 directly. We report that BAF copurified with nuclease-digested mononucleosomes and associated with modified histones in vivo. BAF overexpression significantly reduced global histone H3 acetylation by 18%. In cells that stably overexpressed BAF 3-fold, silencing mark H3-K27-Me1/3 and active marks H4-K16-Ac and H4-Ac5 decreased significantly. Significant increases were also seen for silencing mark H3-K9-Me3, active marks H3-K4-Me2, H3-K9/K14-Ac and H4-K5-Ac and a mark (H3-K79-Me2) associated with both active and silent chromatin. Other increases (H3-S10-P, H3-S28-P and silencing mark H3-K9-Me2) did not reach statistical significance. BAF overexpression also significantly influenced cell cycle distribution. Moreover, BAF associated in vivo with SET/I2PP2A (protein phosphatase 2A inhibitor; blocks H3 dephosphorylation) and G9a (H3-K9 methyltransferase), but showed no detectable association with HDAC1 or HATs. These findings reveal BAF as a novel epigenetic regulator and are discussed in relation to BAF deficiency phenotypes, which include a hereditary progeria syndrome and loss of pluripotency in embryonic stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, Anti-acetyl-Histone H4 (Lys16) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16 & has been published and validated in ChIP, WB, Mplex, PIA, DB, ChIP-seq.
07-327
Anti-acetyl-Histone H4 (Lys5) Antibody, Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
06-946
Anti-hyperacetylated Histone H4 (Penta) Antibody, Use Anti-hyperacetylated Histone H4 (Penta) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, ICC, WB to detect hyperacetylated Histone H4 (Penta) also known as H4 histone family member A.
07-145
Anti-phospho-Histone H3 (Ser28) Antibody, Use Anti-phospho-Histone H3 (Ser28) Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC, DB, PIA, ChIP to detect phospho-Histone H3 (Ser28) also known as H3F3, H3F3B, H3S28P.
05-368
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, Anti-phospho-Ser/Thr-Pro MPM-2 Antibody is a Mouse Monoclonal Antibody for detection of phospho-Ser/Thr-Pro also known as Mitotic protein #2 & has been tested in ELISA, IHC, IP & WB.