EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
Inhibitors of intravesicular acidification protect against Shiga toxin in a pH-independent manner.
PMID 22132807
Abstract
Shiga toxin inhibits protein synthesis after being transported from the cell surface to endosomes and retrogradely through the Golgi apparatus to the endoplasmic reticulum (ER) and into the cytosol. In this study, we have abolished proton gradients across internal membranes in different ways and investigated the effect on the various transport steps of Shiga toxin. Although inhibitors of the proton pump such as bafilomycin A1 and concanamycin A as well as some ionophores and chloroquine all protect against Shiga toxin, they mediate protection by inhibiting different transport steps. For instance, chloroquine protects the cells, although the toxin is transported to the ER. Importantly, our data indicate that proton pump activity is required for efficient endosome-to-Golgi transport of Shiga toxin, although acidification as such does not seem to be required.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9705
Concanamycin A, ≥70% (HPLC)
C46H75NO14
27689
Concanamycin A, from Streptomyces sp., ≥80% (HPLC)
C46H75NO14