EMAIL THIS PAGE TO A FRIEND
Clinical and experimental rheumatology
Therapeutic consequences of crystals in the synovial fluid: a review for clinicians.
PMID 22133072
Abstract
Many crystals may be found in arthritic joints. Rheumatologists are able to diagnose, with a high degree of probability, which crystals induce arthritis in the individual case. A definite diagnosis supported by polarisation microscopy may provide a firm basis for adequate long-term treatment. Recently new insights in the inflammatory processes induced the development of targeted therapies. We review novel developments in crystal-induced arthritis and gout in particular.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
693871
Calcium phosphate, amorphous, nanopowder, <150 nm particle size (BET)