EMAIL THIS PAGE TO A FRIEND
Brain research
The combination of organoselenium compounds and guanosine prevents glutamate-induced oxidative stress in different regions of rat brains.
PMID 22133308
Abstract
This study was designed to investigate the protective effects of the combination of guanosine and 2 organoselenium compounds (ebselen and diphenyl diselenide) against glutamate-induced oxidative stress in different regions of rat brains. Glutamate caused an increase in reactive oxygen species (ROS) generation and a decrease in [(3)H]-glutamate uptake in striatal, cortical, and hippocampal slices. Guanosine, ebselen, and diphenyl diselenide prevented glutamate-induced ROS production in striatal, cortical and hippocampal slices. The combination of guanosine with organoselenium compounds was more effective against glutamate-induced ROS production than the individual compounds alone. Guanosine prevented [(3)H]-glutamate uptake inhibition in striatal, cortical, and hippocampal slices. Thus, protection against the harmful effects of glutamate is possibly due to the combination of the antioxidant properties of organoselenium compounds and the stimulatory effect of guanosine on glutamate uptake. In conclusion, the combination of antioxidants and glutamatergic system modulators could be considered a potential therapy against the prooxidant effects of glutamate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42944
Diphenyl diselenide, purum, ≥97.0% (GC)
C12H10Se2
180629
Diphenyl diselenide, 98%
C12H10Se2