EMAIL THIS PAGE TO A FRIEND
Steroids
Synthesis and biological activities of vitamin D-like inhibitors of CYP24 hydroxylase.
PMID 22133546
Abstract
Selective inhibitors of CYP24A1 represent an important synthetic target in a search for novel vitamin D compounds of therapeutic value. In the present work, we show the synthesis and biological properties of two novel side chain modified 2-methylene-19-nor-1,25(OH)(2)D(3) analogs, the 22-imidazole-1-yl derivative 2 (VIMI) and the 25-N-cyclopropylamine compound 3 (CPA1), which were efficiently prepared in convergent syntheses utilizing the Lythgoe type Horner-Wittig olefination reaction. When tested in a cell-free assay, both compounds were found to be potent competitive inhibitors of CYP24A1, with the cyclopropylamine analog 3 exhibiting an 80-1 selective inhibition of CYP24A1 over CYP27B1. Addition of 3 to a mouse osteoblast culture sustained the level of 1,25(OH)(2)D(3), further demonstrating its effectiveness in CYP24A1 inhibition. Importantly, the in vitro effects on human promyeloid leukemia (HL-60) cell differentiation by 3 were nearly identical to those of 1,25(OH)(2)D(3) and in vivo the compound showed low calcemic activity. Finally, the results of preliminary theoretical studies provide useful insights to rationalize the ability of analog 3 to selectively inhibit the cytochrome P450 isoform CYP24A1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125504
Cyclopropylamine, 98%
C3H7N