EMAIL THIS PAGE TO A FRIEND
Medecine et maladies infectieuses
[Treatment of resistant Gram-positive bacterial infections].
PMID 22133738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9393
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic
C19H17ClN3NaO5S · H2O