EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
The interplay between mutasynthesis and semisynthesis: generation and evaluation of an ansamitocin library.
PMID 22135226
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2836
Ansamitocin P-3 from Actinosynnema pretiosum, ≥90% (HPLC)
C32H43ClN2O9