EMAIL THIS PAGE TO A FRIEND
Health policy (Amsterdam, Netherlands)
From evidence assessments to coverage decisions?: the case example of glinides in Germany.
PMID 22136812
Abstract
In Germany, coverage decisions in the statutory health insurance (SHI) system are based on the principles of evidence-based medicine. Recently, an evidence assessment by the Institute for Quality and Efficiency in Health Care (IQWiG) of the oral antidiabetics of the glinide class showed that their long-term benefit is not proven. Accordingly, the responsible Federal Joint Committee (G-BA) decided to exclude glinides from prescription in the SHI system. This was, however, objected to by the Ministry of Health, which is charged with legal supervision. We use this case to illustrate the path from evidence assessments to coverage decisions in Germany against the background of the latest health reform, which has changed the legal requirements for evidence assessments and the ensuing coverage decisions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3538
Nateglinide, ≥98% (HPLC), solid
C19H27NO3