EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Efficient double glycoconjugation to naturalize high molecular weight disperse dyes.
PMID 22137592
Abstract
Commercially available Disperse Orange 29 (1a) and Disperse Red 1 (2a) were elaborated to glycoconjugated species, following a new version of a previously-described 'naturalisation' procedure. Glutamic acid was chosen to achieve a double glycoconjugation, which is essential to give to the original disperse dye a water solubility suitable for reaching optimal dyeing conditions. UV-vis plot of the 'naturalised' species showed negligible differences when compared to those of the commercial dyes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
344206
Disperse Red 1, Dye content 95 %
C16H18N4O3
11074
Disperse Red 1, analytical standard
C16H18N4O3