EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
Benzyl butyl phthalate induces necrosis by AhR mediation of CYP1B1 expression in human granulosa cells.
PMID 22138065
Abstract
We investigated the signaling pathway of the arylhydrocarbon receptor (AhR) on HO23 cells (immortalized human granulosa cells (hGC)) mediated by benzyl butyl-phthalate (BBP). BBP (1 μM) significantly increased the mRNA and protein levels of AhR, aryl hydrocarbon receptor nuclear translocator (ARNT) and cytochrome-P450 (CYP)1B1 in HO23 cells. Treatment with 3',4'-dimethoxyflavone (3',4'-DMF) or AhR siRNA significantly reduced AhR and CYP1B1, but CYP1A1 was not affected by 3',4'-DMF or AhR siRNA, suggesting that increases in CYP1A1 may not regulated by AhR. BBP induced the AhR fusion protein to localize and accumulate around the nucleus, and AhR heterodimerization with ARNT was observed in the nucleus by immunoprecipitation. Chromatin immunoprecipitation and reporter assays revealed the effect of BBP on CYP1B1, but not CYP1A1. Necrosis was significantly increased in HO23 cells after BBP treatment, and 3',4'-DMF, AhR siRNA or CYP1B1 siRNA knockdown blocked this phenomenon. These data suggest that BBP-induced HO23 cell necrosis is AhR and CYP1B1 dependent.