EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Tertiary alkylamines as nucleophiles in substitution reactions at heteroaromatic halide during the synthesis of the highly potent pirinixic acid derivative 2-(4-chloro-6-(2,3-dimethylphenylamino)pyrimidin-2-ylthio)octanoic acid (YS-121).
PMID 22143571
Abstract
YS-121 [2-(4-chloro-6-(2,3-dimethylphenylamino)pyrimidin-2-ylthio)octanoic acid] is the result of target-oriented structural derivatization of pirinixic acid. It is a potent dual PPARα/γ-agonist, as well as a potent dual 5-LO/mPGES-1-inhibitor. Additionally, recent studies showed an anti-inflammatory efficacy in vivo. Because of its interference with many targets, YS-121 is a promising drug candidate for the treatment of inflammatory diseases. Ongoing preclinical studies will thus necessitate huge amounts of YS-121. To cope with those requirements, we have optimized the synthesis of YS-121. Surprisingly, we isolated and characterized byproducts during the resulting from nucleophilic aromatic substitution reactions by different tertiary alkylamines at a heteroaromatic halide. These amines should actually serve as assisting bases, because of their low nucleophilicity. This astonishing fact was not described in former publications concerning that type of reaction and, therefore, might be useful for further reaction improvement in general. Furthermore, we could develop a proposal for the mechanism of that byproduct formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7081
WY-14643
C14H14ClN3O2S