EMAIL THIS PAGE TO A FRIEND
Indian journal of orthopaedics
Antibiotic-impregnated articulating cement spacer for infected total knee arthroplasty.
PMID 22144747
Abstract
Standard treatment of chronic infected total knee arthroplasty (TKA) is a two-stage revision, the first step being placement of an antibiotic-impregnated cement spacer. Here we describe the results of a new technique (modification of the Goldstien's technique) for intraoperative manufacture of a customized articulating spacer at minimal cost and with relatively good conformity and longevity. Thirty-six infected knees underwent this procedure from June 2002 to May 2007. The technique consists of using the freshened femur and tibia interface as molds wrapped in a tin foil for manufacturing the two components of the spacer with antibiotic-impregnated methyl methycrylate cement. We used the spacer and the femoral component of the trial set of a TKA system to mold them to perfect articulation. We also reinforced the spacer with a K-wire scaffold to prevent fracture of the cement mantle in the last 21 cases. All 36 knees showed excellent results in terms of infection control, mobility, and stability. There was significant improvement in the WOMAC and Knee Society Scores (20 and 39 points respectively). There were two fractures of the spacers in the initial 15 cases that did not have K-wire scaffolding but none in the last 21 that had reinforcement. This technique provides a more conforming spacer, with good range of motion and stability. The reinforcement helps in preventing the fracture of the cement mantle and is cost effective.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF66339254
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF54760259
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF45567434
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF28221274
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF16505636
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF50720925
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF00846413
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF99151881
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF83193925
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF86850029
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF17353425
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF40941097
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF66628996
Tin, foil, 4mm disks, thickness 0.007mm, 97.4%
Sn
GF33080532
Tin, foil, 4mm disks, thickness 0.015mm, 99.75%
Sn
GF29177039
Tin, foil, 4mm disks, thickness 0.02mm, 99.75%
Sn
GF51371232
Tin, foil, 4mm disks, thickness 0.125mm, as rolled, 98.8%
Sn
GF34214464
Tin, foil, 4mm disks, thickness 0.25mm, as rolled, 98.8%
Sn
GF63204106
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Sn
GF04830331
Tin, foil, 50mm disks, thickness 0.0125mm, 97.4%
Sn
GF96864643
Tin, foil, 50mm disks, thickness 0.05mm, as rolled, 98.8%
Sn
GF07913088
Tin, foil, 8mm disks, thickness 0.06mm, as rolled, 99.99+%
Sn
GF18183764
Tin, foil, 8mm disks, thickness 0.1mm, as rolled, 98.8%
Sn
GF36451098
Tin, foil, 8mm disks, thickness 0.65mm, as rolled, 99.99+%
Sn
GF00022699
Tin, foil, light tested, 100x100mm, thickness 0.0125mm, 97.4%
Sn