EMAIL THIS PAGE TO A FRIEND
Acta anaesthesiologica Scandinavica
'Intense inner agitation': an overlooked side effect of droperidol.
PMID 22150523
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3100000
Droperidol, European Pharmacopoeia (EP) Reference Standard
C22H22FN3O2