EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of potent small molecule inhibitors of DYRK1A by structure-based virtual screening and bioassay.
PMID 22154664
Abstract
In this study, six novel dual-specificity tyrosine phosphorylation regulated kinase 1A (DYRK1A) inhibitors with IC(50) values ranging from 1.51 to 88.13 μM were successfully identified through virtual screening and in vitro plus cell based bioassay. Compound 5 with IC(50) value of 1.51 μM is the most potent hit against DYRK1A in vitro, while compound 3 exhibited the most potent activity in cultured cells. The inhibition mechanism was explored by molecular docking approach. This study may provide a start point for further mechanism based study as well as discovery of drug candidate against Down syndrome (DS).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13-110
MBP, Dephosphorylated